Learn More
AdipoGen Betulinic acid (>99%) (5 mg)
SDP
Supplier:  AdipoGen AGCN20415M005
Encompass_Preferred
Chemical. CAS: 472-15-1. Formula: C30H48O3. MW: 456.7. Isolated from Platanus acerifolia bark. Anti-HIV compound. Anticancer compound. Apoptosis inducer. Modulator of NF-kappaB. Potent DNA topoisomerase II inhibitor. Potent proteasome activator. Antimalarial compound.
Catalog No. 50-205-3586