Learn More
Cayman Chemical Betulinic Acid methyl ester, Purity: ≥95%, CAS: 2259-06-5, 1 mg
SDP
Catalog No. 502988610
Encompass_Preferred
A triterpenoid and an esterified version of betulinic acid with diverse biological activities; inhibits HIV-1 reverse transcriptase (IC50 = 11 μM); active against L. braziliensis and T. cruzi (IC50s = 69.9 and 93.3 μM, respectively); induces melanogenesis in (EC50 = 2.5 μM) and inhibits the growth of B16 2F2 melanoma cells in vitro (IC50 = 4.9 μM); cytotoxic to SK-LU-1, HepG2, HeLa, SK-MEL-2, and AGS cells (IC50s = 60.84, 77.43, 80.17, 66.17, and 69.94 μg/ml, respectively);
Catalog No. 50-298-8610 Supplier Cayman Chemical Supplier No. 300661 MG
A triterpenoid and an esterified version of betulinic acid with diverse biological activities; inhibits HIV-1 reverse transcriptase (IC50 = 11 μM); active against L. braziliensis and T. cruzi (IC50s = 69.9 and 93.3 μM, respectively); induces melanogenesis in (EC50 = 2.5 μM) and inhibits the growth of B16 2F2 melanoma cells in vitro (IC50 = 4.9 μM); cytotoxic to SK-LU-1, HepG2, HeLa, SK-MEL-2, and AGS cells (IC50s = 60.84, 77.43, 80.17, 66.17, and 69.94 μg/ml, respectively);