Learn More
Medchemexpress LLC Betulinic acid (standard) | 472-15-1 | MFCD00009619 | 99.2% | 456.70 | C30H48O3 | 50 MG
SDP
Catalog No. 5000243682
Encompass_Preferred
Betulinic acid (analytical standard) is a high-purity pentacyclic triterpenoid supplied for research and analytical applications. Supplied as a 50 mg solid with accompanying certificate of analysis and safety documentation, it is intended for use as a calibration standard and positive control in chromatographic and quantitative assays.
  • High purity suitable for analytical use.
  • Supplied as a solid 50 mg standard.
  • Provided with COA and SDS for traceability.
  • Suitable as a calibration standard or reference material.
  • Applicable for chromatography, method development, and quantitative analysis.
Catalog No. 50-002-43682 Supplier Medchemexpress LLC Supplier No. HY10529R50MG
Betulinic acid (analytical standard) is a high-purity pentacyclic triterpenoid supplied for research and analytical applications. Supplied as a 50 mg solid with accompanying certificate of analysis and safety documentation, it is intended for use as a calibration standard and positive control in chromatographic and quantitative assays.
  • High purity suitable for analytical use.
  • Supplied as a solid 50 mg standard.
  • Provided with COA and SDS for traceability.
  • Suitable as a calibration standard or reference material.
  • Applicable for chromatography, method development, and quantitative analysis.