Learn More
Cayman Chemical Betulinic Aldehyde, Purity: ≥98%, CAS: 13159-28-9, 5 mg
SDP
Catalog No. 502937894
Encompass_Preferred
A triterpenoid with diverse biological activities; active against P. falciparum (IC50 = 6.5 μg/ml) and M. tuberculosis (MIC = 25 μg/ml) in vitro, as well as laboratory and clinical isolates of methicillin-resistant and -sensitive S. aureus (MICs = 128-512 and 8-512 μg/ml, respectively); inhibits dengue virus NS5 RNA-dependent RNA polymerase (IC50 = 6.1 μM) and inhibits dengue virus type 2 replication by 20 and 55% at 5 and 10 μg/ml, respectively; inhibits growth of leukemia, melanoma, neuroblastoma, and normal fibroblast cell lines (IC50s = 1.1-4.9, 9.6-10.6, 7.5-8.8, and 18.5 μM, respectively)
Catalog No. 50-293-7894 Supplier Cayman Chemical Supplier No. 278105 MG
A triterpenoid with diverse biological activities; active against P. falciparum (IC50 = 6.5 μg/ml) and M. tuberculosis (MIC = 25 μg/ml) in vitro, as well as laboratory and clinical isolates of methicillin-resistant and -sensitive S. aureus (MICs = 128-512 and 8-512 μg/ml, respectively); inhibits dengue virus NS5 RNA-dependent RNA polymerase (IC50 = 6.1 μM) and inhibits dengue virus type 2 replication by 20 and 55% at 5 and 10 μg/ml, respectively; inhibits growth of leukemia, melanoma, neuroblastoma, and normal fibroblast cell lines (IC50s = 1.1-4.9, 9.6-10.6, 7.5-8.8, and 18.5 μM, respectively)