Learn More
Cayman Chemical Betulinic Aldehyde oxime, Purity: ≥95%, CAS: 25613-12-1, 50 mg
SDP
Catalog No. 502946447
Encompass_Preferred
A derivative of betulin; active against E. aerogenes, E. coli, E. faecalis, P. aeruginosa, S. aureus, and C. albicans, as well as L. donovani amastigotes, at 50 µM; is cytotoxic to Huh7 hepatocellular carcinoma cells at 50 µM
Catalog No. 50-294-6447 Supplier Cayman Chemical Supplier No. 3302750 MG
A derivative of betulin; active against E. aerogenes, E. coli, E. faecalis, P. aeruginosa, S. aureus, and C. albicans, as well as L. donovani amastigotes, at 50 µM; is cytotoxic to Huh7 hepatocellular carcinoma cells at 50 µM