Learn More
Cayman Chemical Betulinic glycine amide, Purity: ≥95%, CAS: 174740-40-0, 1 mg
SDP
Catalog No. 502946501
Encompass_Preferred
A derivative of betulinic acid; has been used as a precursor in the synthesis of betulinic acid derivatives with anticancer and antiviral activity
Catalog No. 50-294-6501 Supplier Cayman Chemical Supplier No. 330311 MG
A derivative of betulinic acid; has been used as a precursor in the synthesis of betulinic acid derivatives with anticancer and antiviral activity