Learn More
Cayman Chemical Betulinic Acid propargyl ester, Purity: ≥95%, CAS: 1400262-27-2, 10 mg
SDP
Catalog No. 502946883
Encompass_Preferred
An alkyne derivative of betulinic acid; has been used in click chemistry reactions for the synthesis of cytotoxic anticancer betulinic acid derivatives
Catalog No. 50-294-6883 Supplier Cayman Chemical Supplier No. 3303010 MG
An alkyne derivative of betulinic acid; has been used in click chemistry reactions for the synthesis of cytotoxic anticancer betulinic acid derivatives