Learn More
Cayman Chemical Betulonaldehyde 10mg
SDP
Catalog No. 502949769
Encompass_Preferred
A pentacyclic triterpenoid; active against P. falciparum (IC50 = 3.36 UG/ml); cytotoxic to NCI H187 and Vero cells (IC50s = 19.23 and 17.09 UG/ml, respectively); reduces TPA-induced EAr edema in mice at 1 mg/ear; has also been used a precursor in the semisynthesis of C-2 and C-28 betulonic aldehyde derivatives
Catalog No. 50-294-9769 Supplier Cayman Chemical Supplier No. 3304210 MG
A pentacyclic triterpenoid; active against P. falciparum (IC50 = 3.36 UG/ml); cytotoxic to NCI H187 and Vero cells (IC50s = 19.23 and 17.09 UG/ml, respectively); reduces TPA-induced EAr edema in mice at 1 mg/ear; has also been used a precursor in the semisynthesis of C-2 and C-28 betulonic aldehyde derivatives