Learn More
Cayman Chemical BetulonIc AcId 10mg
SDP
Catalog No. 502988362
Encompass_Preferred
A triterpenoid; active against P. falciparum W2 clone (IC50 = 10.01 µM), as well as L. infantum and L. donovani promastigotes at 50 µM; inhibits replication of HSV-I, echovirus 6, and the H7N1 strain of influenza A (EC50s = 0.9, 73.32, and 5.7 µM, respectively); inhibits proliferation of MGC803, Bcap-37, MCF-7, PC3, and NIH3T3 cancer cells by 68.1, 44.9, 56.1, 52.4, and 22.1%, respectively, at 20 µM
Catalog No. 50-298-8362 Supplier Cayman Chemical Supplier No. 2976510 MG
A triterpenoid; active against P. falciparum W2 clone (IC50 = 10.01 µM), as well as L. infantum and L. donovani promastigotes at 50 µM; inhibits replication of HSV-I, echovirus 6, and the H7N1 strain of influenza A (EC50s = 0.9, 73.32, and 5.7 µM, respectively); inhibits proliferation of MGC803, Bcap-37, MCF-7, PC3, and NIH3T3 cancer cells by 68.1, 44.9, 56.1, 52.4, and 22.1%, respectively, at 20 µM