Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Betulonic acid | 4481-62-3 | MFCD00069004 | 454.70 | 100 MG
SDP

Catalog No. 5000415489
Encompass_Preferred

Betulonic acid is a naturally occurring triterpene found in many plants. It exhibits anti-tumor, anti-inflammatory, antiparasitic, and antiviral (HSV-1) activities.

  • Synonyms include Betunolic acid, Liquidambaric acid, and (+)-Betulonic acid.
  • Purity of 98.77%.
  • Soluble in DMSO.
  • Demonstrates anti-tumor activity.
  • Demonstrates anti-inflammatory activity.
  • Demonstrates antiparasitic activity.
  • Demonstrates anti-viral (HSV-1) activity.

Catalog No. 50-004-15489 Supplier Medchemexpress LLC Supplier No. HYN1451100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Betulonic acid is a naturally occurring triterpene found in many plants. It exhibits anti-tumor, anti-inflammatory, antiparasitic, and antiviral (HSV-1) activities.

  • Synonyms include Betunolic acid, Liquidambaric acid, and (+)-Betulonic acid.
  • Purity of 98.77%.
  • Soluble in DMSO.
  • Demonstrates anti-tumor activity.
  • Demonstrates anti-inflammatory activity.
  • Demonstrates antiparasitic activity.
  • Demonstrates anti-viral (HSV-1) activity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.