Learn More
TARGETMOL CHEMICALS INC Betulonic acid 50MG
SDP
Catalog No. 502876061
Encompass_Preferred
Also available in 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. 1. Betulonic acid (Liquidambaric acid) amide stimulated the regenerative response in hepatocytes under conditions of combined toxic exposure and promoted recovery of their qualitative and quantitative characteristics, which was accompanied by a significant decrease in the severity of liver fibrosis and the absence of cirrhotic transformation of the liver. 2. Betulonic acid derivatives have a promising cytostatic activity in vitro and could be used as potential leads for the development of new type of anti-cancer agents. 3. Betulonic acid has antiholestatic effects in mice. Purity 99.7%
Catalog No. 50-287-6061 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S001850MG
Add to Cart
Add to Cart
Also available in 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. 1. Betulonic acid (Liquidambaric acid) amide stimulated the regenerative response in hepatocytes under conditions of combined toxic exposure and promoted recovery of their qualitative and quantitative characteristics, which was accompanied by a significant decrease in the severity of liver fibrosis and the absence of cirrhotic transformation of the liver. 2. Betulonic acid derivatives have a promising cytostatic activity in vitro and could be used as potential leads for the development of new type of anti-cancer agents. 3. Betulonic acid has antiholestatic effects in mice. Purity 99.7%