Learn More
AVANTOR MASTERFLEX (VWR LLC) Bev-A-Line B-5 V Tubing, 1/8""ID x 1/4""OD, 50 ft/pack
Catalog No. NC0320866
Encompass
Applications: Safe for food, beverage, drug, lab, and general use¿can withstand higher temperatures than Bev-A-Line IV tubing. Safe for most alcohols; avoid strong solvents. Characteristics: Polyethylene liner cross-linked to ethyl vinyl acetate shell. No plasticizers. Nontoxic and nonhemolytic. Needs no connectors¿heat-bondable. Translucent. Certification: Resin is FDA-compliant (liner: 21 CFR 177.1520; shell 21 CFR 177.1350). Meets USP Class VI, USDA, and National Formulary requirements. Temperature range: ¿60 to 200¿F (¿51 to 93¿C) Sterilize: With autoclave, ethylene oxide, or dry heat.
Catalog No. NC0320866 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0649112
Applications: Safe for food, beverage, drug, lab, and general use¿can withstand higher temperatures than Bev-A-Line IV tubing. Safe for most alcohols; avoid strong solvents. Characteristics: Polyethylene liner cross-linked to ethyl vinyl acetate shell. No plasticizers. Nontoxic and nonhemolytic. Needs no connectors¿heat-bondable. Translucent. Certification: Resin is FDA-compliant (liner: 21 CFR 177.1520; shell 21 CFR 177.1350). Meets USP Class VI, USDA, and National Formulary requirements. Temperature range: ¿60 to 200¿F (¿51 to 93¿C) Sterilize: With autoclave, ethylene oxide, or dry heat.