Learn More
McMaster-Carr Wet-Food/Beverage Hose with Abrasion-Resistant Cover
Supplier:  McMaster-Carr 54975K312FT
Encompass
Wet-Food/Beverage Hose with Abrasion-Resistant Cover with 316 Stainless Steel Quick-Clamp Fittings, 1-1/2" ID, 2-3/16" OD
Catalog No. NC2894099