Learn More
LAB BIOREAGENTS Bevacizumab Elisa Kit
Supplier:  LAB BIOREAGENTS BEK011132
Encompass
Bevacizumab ELISA Kit is a sandwich enzyme-linked immunoassay for the quantitative determination of Bevacizumab with high sensitivity and specificity in serum and plasma.Kit PerformanceSensitivity: 25 ng/mLAssay Range: 25 – 2000 ng/mLCross-reactivity: There is no cross reaction with native serum immunoglobulinsRecovery rate: 80 – 120% with normal human serum samples with known concentrations.Precision: Intra-Assay: CV < 20%; Inter-Assay: CV < 20%
Catalog No. 50-195-9285