Learn More
Genscript Corporation Antibody Kit Other Products Immunodetection 1/EA
Supplier:  Genscript Corporation L00969
Encompass
The Bevacizumab ELISA kit is a validated tool for quantifying both Bevacizumab and its biosimilars in biological matrices facilitating drug research and development. GenScripts Bevacizumab Pharmacokinetic ELISA Kit has been comprehensively developed and validated for quantitative measurement of Bevacizumab in cynomolgus monkey serum and plasma based on the ICH M10 and the FDA bioanalytical method validation guidance for industry.
Catalog No. 50-252-9976