Learn More
Selleck Chemical LLC Bevantolol hydrochloride S5042-25mg
SDP
Catalog No. 503112454
Encompass_Preferred
Bevantolol hydrochloride (NC-1400) is a cardioselective beta adrenoceptor antagonist devoid of intrinsic sympathomimetic activity and with weak membrane-stabilizing and local anesthetic properties
Catalog No. 50-311-2454 Supplier Selleck Chemical LLC Supplier No. S504225MG
Bevantolol hydrochloride (NC-1400) is a cardioselective beta adrenoceptor antagonist devoid of intrinsic sympathomimetic activity and with weak membrane-stabilizing and local anesthetic properties