Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bevantolol hydrochloride | 42864-78-8 | 10 MG
SDP

Catalog No. 5000234103
Encompass_Preferred

Bevantolol hydrochloride is a beta-1 and alpha-1 adrenergic receptor antagonist supplied as an analytical standard for laboratory research in adrenergic pharmacology.

  • Selective β1 and α1 adrenergic receptor antagonist.
  • Supplied as an analytical standard for research use.
  • CAS number 42864-78-8.
  • Molecular formula C20H28ClNO4; molecular weight 381.89 g/mol.
  • Available in small milligram pack sizes, including 10 MG for analytical applications.

Catalog No. 50-002-34103 Supplier Medchemexpress LLC Supplier No. HY12118610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bevantolol hydrochloride is a beta-1 and alpha-1 adrenergic receptor antagonist supplied as an analytical standard for laboratory research in adrenergic pharmacology.

  • Selective β1 and α1 adrenergic receptor antagonist.
  • Supplied as an analytical standard for research use.
  • CAS number 42864-78-8.
  • Molecular formula C20H28ClNO4; molecular weight 381.89 g/mol.
  • Available in small milligram pack sizes, including 10 MG for analytical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.