Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bevantolol hydrochloride | 42864-78-8 | 99.0% | 1 ML
SDP

Catalog No. 5000231690
Encompass_Preferred

Bevantolol hydrochloride is a selective β1 and α1-adrenergic receptor antagonist supplied as the hydrochloride salt for research and analytical use. It is commonly used as a reference standard in pharmacology and receptor-binding studies.

  • Selective β1 and α1-adrenergic receptor antagonist
  • Supplied as the hydrochloride salt suitable for analytical use
  • Molecular weight 381.89 g/mol
  • Chemical formula C20H28ClNO4
  • Typical purity around 98.98% as reported by the manufacturer
  • Available as solid reference standards; solution formulations may be offered by suppliers

Catalog No. 50-002-31690 Supplier Medchemexpress LLC Supplier No. HY12118610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bevantolol hydrochloride is a selective β1 and α1-adrenergic receptor antagonist supplied as the hydrochloride salt for research and analytical use. It is commonly used as a reference standard in pharmacology and receptor-binding studies.

  • Selective β1 and α1-adrenergic receptor antagonist
  • Supplied as the hydrochloride salt suitable for analytical use
  • Molecular weight 381.89 g/mol
  • Chemical formula C20H28ClNO4
  • Typical purity around 98.98% as reported by the manufacturer
  • Available as solid reference standards; solution formulations may be offered by suppliers

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.