Learn More
Medchemexpress LLC Bevantolol hydrochloride | 42864-78-8 | 99.0% | 5 MG
SDP
Catalog No. 5000233503
Encompass_Preferred
Bevantolol hydrochloride is a selective β1 and α1-adrenergic receptor antagonist and calcium channel antagonist provided as a research-grade analytical standard for pharmacology and signaling studies.
  • Selective β1 and α1-adrenergic receptor antagonist.
  • Also acts as a calcium channel antagonist.
  • Available in small milligram pack sizes suitable for research.
  • Soluble in DMSO and water; formulation guidance provided.
  • Store sealed at 4°C; solvent storage recommendations provided.
Catalog No. 50-002-33503 Supplier Medchemexpress LLC Supplier No. HY1211865MG
Bevantolol hydrochloride is a selective β1 and α1-adrenergic receptor antagonist and calcium channel antagonist provided as a research-grade analytical standard for pharmacology and signaling studies.
  • Selective β1 and α1-adrenergic receptor antagonist.
  • Also acts as a calcium channel antagonist.
  • Available in small milligram pack sizes suitable for research.
  • Soluble in DMSO and water; formulation guidance provided.
  • Store sealed at 4°C; solvent storage recommendations provided.