Learn More
MSC SPI Universal Bevel Protractor, Hardened Stainless Steel, 12 in. Blade, 1 ea.
Catalog No. 5000511412
Encompass
These bevel protractor blades and acute angle attachments are designed for use with a universal bevel protractor. Engineered for precision dimensional measurement, these components are constructed from hardened stainless steel to ensure durability and accuracy in demanding metalworking environments.
  • Hardened stainless steel construction
  • Includes 12 in. and 6 in. blades
  • Includes reference square
  • Supplied in a fitted case
Catalog No. 50-005-11412 Supplier MSC Supplier No. 01719046
These bevel protractor blades and acute angle attachments are designed for use with a universal bevel protractor. Engineered for precision dimensional measurement, these components are constructed from hardened stainless steel to ensure durability and accuracy in demanding metalworking environments.
  • Hardened stainless steel construction
  • Includes 12 in. and 6 in. blades
  • Includes reference square
  • Supplied in a fitted case