Learn More
Med Vet International Beveled Double Frosted Slides, One End Both Sides, 144/Box, 10 Boxes/Case
Catalog No. 503043143
Encompass_Preferred
These beveled edge slides are made of water white glass and have clipped corners (1mm). 75x25mm (3?x1?) approx. 1mm thick. 144/GR 10GR/CS
Catalog No. 50-304-3143 Supplier Med Vet International Supplier No. 8207CS
These beveled edge slides are made of water white glass and have clipped corners (1mm). 75x25mm (3?x1?) approx. 1mm thick. 144/GR 10GR/CS