Learn More
Med Vet International Beveled Plain Slides, 144/Box, 10 Boxes/Case
Catalog No. 503043896
Encompass_Preferred
These beveled edge slides are made of water white glass and have clipped corners (1mm). 75x25mm (3?x1?) approx. 1mm thick. 72pcs per box, 144/Box, 10 Boxes/Case
Catalog No. 50-304-3896 Supplier Med Vet International Supplier No. 8201CS
These beveled edge slides are made of water white glass and have clipped corners (1mm). 75x25mm (3?x1?) approx. 1mm thick. 72pcs per box, 144/Box, 10 Boxes/Case