Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bevenopran | 676500-67-7 | MFCD26967979 | 99.8% | 386.44 g/mol | C20H26N4O4 | 50 MG
SDP

Catalog No. 5000215784
Encompass_Preferred

Bevenopran is a peripherally acting μ-opioid receptor antagonist, with additional activity at δ-opioid receptors, investigated for treatment of opioid-induced bowel dysfunction. It is supplied as a high-purity research compound for use in in vitro and in vivo pharmacology studies.

  • Peripherally acting μ-opioid receptor antagonist with δ-opioid receptor activity.
  • Intended for research use in studies of opioid-induced bowel dysfunction and related pharmacology.
  • High purity (99.82%) suitable for in vitro and in vivo experiments.
  • Molecular weight 386.44 g/mol; chemical formula C20H26N4O4.
  • CAS number 676500-67-7 for unambiguous substance identification.
  • Available as solid quantities (5 mg-200 mg) and as 10 mM solution in DMSO.

Catalog No. 50-002-15784 Supplier Medchemexpress LLC Supplier No. HY10012250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bevenopran is a peripherally acting μ-opioid receptor antagonist, with additional activity at δ-opioid receptors, investigated for treatment of opioid-induced bowel dysfunction. It is supplied as a high-purity research compound for use in in vitro and in vivo pharmacology studies.

  • Peripherally acting μ-opioid receptor antagonist with δ-opioid receptor activity.
  • Intended for research use in studies of opioid-induced bowel dysfunction and related pharmacology.
  • High purity (99.82%) suitable for in vitro and in vivo experiments.
  • Molecular weight 386.44 g/mol; chemical formula C20H26N4O4.
  • CAS number 676500-67-7 for unambiguous substance identification.
  • Available as solid quantities (5 mg-200 mg) and as 10 mM solution in DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.