Learn More
Waters Corp Waters Beverage Analysis Standards Solution, 5 Standards, Liquid, 100 mL
Catalog No. 50201500 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This standards solution is designed for the analysis of soft drink formulations to determine sweeteners and preservatives. It is a liquid formulation containing five specific standards in a 100 mL volume, making it suitable for laboratory use in food applications.
  • Designed for the analysis of soft drink formulations.
  • Determines sweeteners and preservatives.
  • Liquid formulation.
  • Contains acesulfame-K (150 mg/L), saccharin (100 mg/L), caffeine (100 mg/L), benzoate (200 mg/L), and sorbate (100 mg/L).
  • Can be stored at room temperature.
  • Has a shelf life of 12 months.
  • Offers a shelf life of 1 week upon reconstitution.
  • Ships in ambient conditions.
  • Suitable for LC system types.
  • Used for calibration.
  • Applicable in food analysis.
  • Provided as 1 unit per package.
Catalog No. 50-201-500 Supplier Waters Corp Supplier No. 186006008
This standards solution is designed for the analysis of soft drink formulations to determine sweeteners and preservatives. It is a liquid formulation containing five specific standards in a 100 mL volume, making it suitable for laboratory use in food applications.
  • Designed for the analysis of soft drink formulations.
  • Determines sweeteners and preservatives.
  • Liquid formulation.
  • Contains acesulfame-K (150 mg/L), saccharin (100 mg/L), caffeine (100 mg/L), benzoate (200 mg/L), and sorbate (100 mg/L).
  • Can be stored at room temperature.
  • Has a shelf life of 12 months.
  • Offers a shelf life of 1 week upon reconstitution.
  • Ships in ambient conditions.
  • Suitable for LC system types.
  • Used for calibration.
  • Applicable in food analysis.
  • Provided as 1 unit per package.