Learn More
Waters Corp Waters Beverage Analysis Kit, Quantifies 6 Additives, Soft Drinks, Eco-friendly Reagents
Catalog No. 50465913 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This comprehensive, easy-to-use kit is designed for the rapid quantification of six common additives (acesulfame-K, saccharin, caffeine, benzoate, sorbate, and aspartame) in soft drink formulations. It contains sufficient reagents for approximately one month of typical analysis usage.
  • Rapid quantification of six common additives in soft drink formulations.
  • Comprehensive and easy-to-use kit.
  • Shortens analyst preparation time.
  • Reduces lab-to-lab or analyst-to-analyte variability.
  • Eco-friendly, ethanol-based cleaning agents with low toxicity and inexpensive disposal.
  • Boosts laboratory productivity.
  • Enhances data quality.
  • Reduces costs.
  • Improves product consistency.
Catalog No. 50-465-913 Supplier Waters Corp Supplier No. 176002534
This comprehensive, easy-to-use kit is designed for the rapid quantification of six common additives (acesulfame-K, saccharin, caffeine, benzoate, sorbate, and aspartame) in soft drink formulations. It contains sufficient reagents for approximately one month of typical analysis usage.
  • Rapid quantification of six common additives in soft drink formulations.
  • Comprehensive and easy-to-use kit.
  • Shortens analyst preparation time.
  • Reduces lab-to-lab or analyst-to-analyte variability.
  • Eco-friendly, ethanol-based cleaning agents with low toxicity and inexpensive disposal.
  • Boosts laboratory productivity.
  • Enhances data quality.
  • Reduces costs.
  • Improves product consistency.