Learn More
Waters Corp Waters Corporation Mobile Phase Reagent, Beverage Analysis, Acetate Buffer Solution, 1 L Refill
Catalog No. 50201498 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Beverage Analysis Mobile-Phase Reagent is a ready-to-use acetate buffer solution (1 L refill) for analyzing soft drink formulations. Designed for non-chemists, it simplifies the rapid analysis of sweeteners and preservatives, including acesulfame-K, saccharin, caffeine, benzoate, sorbate, and aspartame, providing results in 7-10 minutes with LC methods.
  • Ready-to-use acetate buffer solution.
  • Designed for use by non-chemists.
  • Facilitates rapid analysis of six additives: acesulfame-K, saccharin, caffeine, benzoate, sorbate, and aspartame.
  • Allows for results in as little as 7 minutes (with UPLC) or 10 minutes (with HPLC).
Catalog No. 50-201-498 Supplier Waters Corp Supplier No. 186006006
The Beverage Analysis Mobile-Phase Reagent is a ready-to-use acetate buffer solution (1 L refill) for analyzing soft drink formulations. Designed for non-chemists, it simplifies the rapid analysis of sweeteners and preservatives, including acesulfame-K, saccharin, caffeine, benzoate, sorbate, and aspartame, providing results in 7-10 minutes with LC methods.
  • Ready-to-use acetate buffer solution.
  • Designed for use by non-chemists.
  • Facilitates rapid analysis of six additives: acesulfame-K, saccharin, caffeine, benzoate, sorbate, and aspartame.
  • Allows for results in as little as 7 minutes (with UPLC) or 10 minutes (with HPLC).