Learn More
Waters Corp Waters Corporation Reagent, 1 L, Ethanol, Beverage Analysis
Catalog No. 50201499 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 1 L ready-to-use ethanol-based wash reagent is designed for the analysis of soft drink formulations, specifically for sweeteners and preservatives. It offers a streamlined, easy-to-follow methodology. Purified and quality-checked by liquid chromatography, it ensures consistent results, is pre-formulated for convenience, and is pH-balanced for most acidic and basic chemicals.
  • Used for the analysis of soft drink formulations for sweeteners and preservatives.
  • 1 L ready-to-use ethanol solution.
  • Based on ecologically friendly solvents.
  • Offers a streamlined, easy-to-follow methodology.
  • Purified and quality checked by liquid chromatography.
  • Pre-formulated for convenience.
  • pH-balanced for most acidic and basic chemicals.
  • Component of the Beverage Analysis Kit.
Catalog No. 50-201-499 Supplier Waters Corp Supplier No. 186006007
This 1 L ready-to-use ethanol-based wash reagent is designed for the analysis of soft drink formulations, specifically for sweeteners and preservatives. It offers a streamlined, easy-to-follow methodology. Purified and quality-checked by liquid chromatography, it ensures consistent results, is pre-formulated for convenience, and is pH-balanced for most acidic and basic chemicals.
  • Used for the analysis of soft drink formulations for sweeteners and preservatives.
  • 1 L ready-to-use ethanol solution.
  • Based on ecologically friendly solvents.
  • Offers a streamlined, easy-to-follow methodology.
  • Purified and quality checked by liquid chromatography.
  • Pre-formulated for convenience.
  • pH-balanced for most acidic and basic chemicals.
  • Component of the Beverage Analysis Kit.