Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Standard, 175 mg/L Caffeine, 500 mL, 15% Denatured Ethanol

Catalog No. 50466291 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This product is a calibration standard specifically designed for beverage analysis, featuring a precise concentration of caffeine in an ethanol and MilliQ water solution.

  • Caffeine concentration of 175 mg/L ± 1%.
  • Volume of 500 mL.
  • Solvent composed of 15% denatured ethanol and 85% MilliQ water.
  • Features reversed phase separation mode.
  • Designed for UV detection method.
  • Presented in a liquid formulation.
  • Recommended storage at room temperature.
  • Compatible with LC system types.
  • Provides quantitative results.
  • Offers a shelf life of 24 months.
  • Ships under ambient conditions.
  • Suitable for food applications.
  • Supplied as 1 unit per package.

Catalog No. 50-466-291 Supplier Waters Corp Supplier No. 186006351
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is a calibration standard specifically designed for beverage analysis, featuring a precise concentration of caffeine in an ethanol and MilliQ water solution.

  • Caffeine concentration of 175 mg/L ± 1%.
  • Volume of 500 mL.
  • Solvent composed of 15% denatured ethanol and 85% MilliQ water.
  • Features reversed phase separation mode.
  • Designed for UV detection method.
  • Presented in a liquid formulation.
  • Recommended storage at room temperature.
  • Compatible with LC system types.
  • Provides quantitative results.
  • Offers a shelf life of 24 months.
  • Ships under ambient conditions.
  • Suitable for food applications.
  • Supplied as 1 unit per package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.