Learn More
Medchemexpress LLC HY-N0842 5mg Medchemexpress, Bevirimat CAS:174022-42-5 Purity:>98%
SDP
Catalog No. 501933744
Encompass_Preferred
Medchemexpress, HY-N0842 5mg Bevirimat CAS:174022-42-5 Bevirimat (PA-457; MPC-4326; YK FH312) is an anti-HIV drug derived from a betulinic acid-like compound; is believed to inhibit HIV by a novel mechanism, so-called maturation inhibition. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3744 Supplier Medchemexpress LLC Supplier No. HYN08425MG
Medchemexpress, HY-N0842 5mg Bevirimat CAS:174022-42-5 Bevirimat (PA-457; MPC-4326; YK FH312) is an anti-HIV drug derived from a betulinic acid-like compound; is believed to inhibit HIV by a novel mechanism, so-called maturation inhibition. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.