Learn More
Medchemexpress LLC Bevurogant | 1817773-66-2 | 99.5% | 532.62 g·mol⁻¹ | C26H28N8O3S | 5 MG
SDP
Catalog No. 5000262314
Encompass_Preferred
Bevurogant is a small-molecule antagonist of the retinoid-related orphan receptor gamma t (RORγt) supplied for laboratory research into immune signaling and chronic inflammatory disease models. It is provided as a solid and as a DMSO solution for in vitro and preclinical studies.
  • Small-molecule RORγt antagonist for modulation of Th17-related pathways.
  • High purity: 99.54% suitable for research assays.
  • Chemical formula C26H28N8O3S; molecular weight 532.62 g·mol⁻¹.
  • Available as solid and 10 mM solution in DMSO for flexible use.
  • Intended for in vitro and preclinical research on chronic inflammatory disease models.
Catalog No. 50-002-62314 Supplier Medchemexpress LLC Supplier No. HY1328105MG
Bevurogant is a small-molecule antagonist of the retinoid-related orphan receptor gamma t (RORγt) supplied for laboratory research into immune signaling and chronic inflammatory disease models. It is provided as a solid and as a DMSO solution for in vitro and preclinical studies.
  • Small-molecule RORγt antagonist for modulation of Th17-related pathways.
  • High purity: 99.54% suitable for research assays.
  • Chemical formula C26H28N8O3S; molecular weight 532.62 g·mol⁻¹.
  • Available as solid and 10 mM solution in DMSO for flexible use.
  • Intended for in vitro and preclinical research on chronic inflammatory disease models.