Learn More
Cayman Chemical Bexarotene-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2182068-00-2, 1 mg
SDP
Catalog No. 502964483
Encompass_Preferred
An internal standard for the quantification of bexarotene by GC- or LC-MS
Catalog No. 50-296-4483 Supplier Cayman Chemical Supplier No. 226101 MG
An internal standard for the quantification of bexarotene by GC- or LC-MS