Learn More
Apexbio Technology LLC Bexarotene 153559-49-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061786
Encompass_Preferred
Bexarotene is a synthetic selective agonist for retinoid X receptors (RXRs) a subclass of nuclear hormone receptors involved in the regulation of gene transcription Upon binding to RXRs Bexarotene modulates the expression of target genes associated with cell differentiation proliferation and apoptosis Due to this mechanism Bexarotene is commonly employed in research investigating tumor growth inhibition cellular differentiation pathways and apoptosis induction Researchers also frequently utilize this retinoid derivative to elucidate RXR-mediated signaling pathways in oncology and to explore potential therapeutic applications for various malignancies
Catalog No. 50-000-61786 Supplier Apexbio Technology LLC Supplier No. A83805.1
Bexarotene is a synthetic selective agonist for retinoid X receptors (RXRs) a subclass of nuclear hormone receptors involved in the regulation of gene transcription Upon binding to RXRs Bexarotene modulates the expression of target genes associated with cell differentiation proliferation and apoptosis Due to this mechanism Bexarotene is commonly employed in research investigating tumor growth inhibition cellular differentiation pathways and apoptosis induction Researchers also frequently utilize this retinoid derivative to elucidate RXR-mediated signaling pathways in oncology and to explore potential therapeutic applications for various malignancies