Learn More
Medchemexpress LLC Bexarotene-d4 | 2182068-00-2 | 98.5% | C24H24D4O2 | 1 MG
SDP
Catalog No. 5000353466
Encompass_Preferred
Bexarotene-d4 is a deuterium-labeled compound, serving as a selective retinoid X receptors (RXR) agonist. It is used as a tracer or an internal standard for quantitative analysis in various methods.
  • Used for treating cutaneous T-cell lymphoma
  • Suitable as a tracer for quantitative analysis
  • Can be used as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, or LC-MS methods
Catalog No. 50-003-53466 Supplier Medchemexpress LLC Supplier No. HY14171S1MG
Bexarotene-d4 is a deuterium-labeled compound, serving as a selective retinoid X receptors (RXR) agonist. It is used as a tracer or an internal standard for quantitative analysis in various methods.
  • Used for treating cutaneous T-cell lymphoma
  • Suitable as a tracer for quantitative analysis
  • Can be used as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, or LC-MS methods