Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bexarotene-d4 | 2182068-00-2 | 98.5% | C24H24D4O2 | 10MG
SDP

Catalog No. 5000261435
Encompass_Preferred

Bexarotene-d4 is a deuterium-labeled isotopologue of bexarotene supplied as a research-grade analytical standard for use as an internal standard and tracer in quantitative mass spectrometry and pharmacokinetic studies involving retinoid X receptor ligands. The material is provided as a high-purity solid suitable for analytical applications.

  • Deuterium-labeled tracer for accurate mass spectrometry quantitation.
  • High purity (98.5%) suitable for analytical standards.
  • Molecular formula C24H24D4O2 and molecular weight ~352.50 g/mol.
  • Available in small vial sizes convenient for laboratory use.
  • For research use only; not for human or clinical applications.

Catalog No. 50-002-61435 Supplier Medchemexpress LLC Supplier No. HY14171S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bexarotene-d4 is a deuterium-labeled isotopologue of bexarotene supplied as a research-grade analytical standard for use as an internal standard and tracer in quantitative mass spectrometry and pharmacokinetic studies involving retinoid X receptor ligands. The material is provided as a high-purity solid suitable for analytical applications.

  • Deuterium-labeled tracer for accurate mass spectrometry quantitation.
  • High purity (98.5%) suitable for analytical standards.
  • Molecular formula C24H24D4O2 and molecular weight ~352.50 g/mol.
  • Available in small vial sizes convenient for laboratory use.
  • For research use only; not for human or clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.