Learn More
Medchemexpress LLC Bexarotene derivative | 2140911-49-3 | 99.2% | 518.73 | C32H46N4O2 | 5 MG
SDP
Catalog No. 5000221572
Encompass_Preferred
OAB-14 is a small-molecule bexarotene derivative used as a research probe to investigate Alzheimer's disease-related pathology. It has been reported to increase β-amyloid clearance in APP/PS1 models, improve endosomal-autophagic-lysosomal function, and restore autophagy flux through modulation of the AMPK/mTOR pathway. Supplied as a high-purity solid, it is intended for preclinical research applications with recommended frozen storage for stability.
  • Increases β-amyloid clearance in APP/PS1 models.
  • Modulates AMPK/mTOR signaling to restore autophagy flux.
  • Ameliorates endosomal-autophagic-lysosomal pathway dysfunction.
  • Available in small-scale milligram quantities for preclinical studies.
  • High reported purity suitable for research applications.
  • Stable under recommended frozen storage for long-term use.
Catalog No. 50-002-21572 Supplier Medchemexpress LLC Supplier No. HY1399735MG
OAB-14 is a small-molecule bexarotene derivative used as a research probe to investigate Alzheimer's disease-related pathology. It has been reported to increase β-amyloid clearance in APP/PS1 models, improve endosomal-autophagic-lysosomal function, and restore autophagy flux through modulation of the AMPK/mTOR pathway. Supplied as a high-purity solid, it is intended for preclinical research applications with recommended frozen storage for stability.
  • Increases β-amyloid clearance in APP/PS1 models.
  • Modulates AMPK/mTOR signaling to restore autophagy flux.
  • Ameliorates endosomal-autophagic-lysosomal pathway dysfunction.
  • Available in small-scale milligram quantities for preclinical studies.
  • High reported purity suitable for research applications.
  • Stable under recommended frozen storage for long-term use.