Learn More
Medchemexpress LLC HY-50673 500mg , BEZ235 Dactolisib;NVP-BEZ235 CAS:915019-65-7 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY50673500MG
Encompass_Preferred
Medchemexpress, HY-50673 500mg BEZ235 Dactolisib;NVP-BEZ235 CAS:915019-65-7 Formula:C30H23N5O IC50: 4 nM (p110α), 5 nM (p110γ), 7 nM (p110δ), 75 nM (p110β), 20.7 nM (mTOR) mTORC1, mTORC2 Purity:98% BEZ235 is a dual pan-class I PI3K and mTOR kinase inhibitor with IC 50 of 4 nM/5 nM/7 nM/75 nM, and 20.7 nM for p110α / p110γ / p110δ / p110β and mTOR , respectively. BEZ235 inhibits mTORC1 and mTORC2 . Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2346