Learn More
Apexbio Technology LLC BEZ235 (NVP-BEZ235), 500mg. Cas: 915019-65-7 MFCD: MFCD10565944. PI3K/mTOR inhibitor,ATP-competitve
SDP
Supplier:  Apexbio Technology LLC A8246500
Encompass_Preferred
BEZ235 is an imidazoquinoline derivative inhibiting both PI3K and mTOR kinases with low nanomolar IC50s. It was well tolerated in preclinical animal studies as well as in clinical trials with manageable gastrointestinal side-effects[1, 2]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5090