Learn More
Medchemexpress LLC HY-B0637 100mg , Bezafibrate BM15075 CAS:41859-67-0 Purity:98%
SDP
Catalog No. 501493191
Encompass_Preferred
Medchemexpress, HY-B0637 100mg Bezafibrate BM15075 CAS:41859-67-0 Formula:C19H20ClNO4 EC50: 90 μM (Murine PPARα), 55 μM (Murine PPARγ), 110 μM (Murine PPARδ), 50 μM (Human PPARα), 60 μM (Human PPARγ), 20 μM (Human PPARδ) Purity:98% Bezafibrate is an agonist of PPAR , with EC 50 s of 50 μM, 60 μM, 20 μM for human PPARα , PPARγ and PPARδ , and 90 μM, 55 μM, 110 μM for murine PPARα , PPARγ and PPARδ , respectively; Bezafibrate is used as an hypolipidemic agent. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3191 Supplier Medchemexpress LLC Supplier No. HYB0637100MG
Add to Cart
Add to Cart
Medchemexpress, HY-B0637 100mg Bezafibrate BM15075 CAS:41859-67-0 Formula:C19H20ClNO4 EC50: 90 μM (Murine PPARα), 55 μM (Murine PPARγ), 110 μM (Murine PPARδ), 50 μM (Human PPARα), 60 μM (Human PPARγ), 20 μM (Human PPARδ) Purity:98% Bezafibrate is an agonist of PPAR , with EC 50 s of 50 μM, 60 μM, 20 μM for human PPARα , PPARγ and PPARδ , and 90 μM, 55 μM, 110 μM for murine PPARα , PPARγ and PPARδ , respectively; Bezafibrate is used as an hypolipidemic agent. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.