Learn More
Cayman Chemical BezafIbrate-d4 1mg
SDP
Catalog No. 502940323
Encompass_Preferred
An internal standard for the quantification of bezafibrate by GC- or LC-MS
Catalog No. 50-294-0323 Supplier Cayman Chemical Supplier No. 285261 MG
Add to Cart
Add to Cart
An internal standard for the quantification of bezafibrate by GC- or LC-MS