Learn More
Medchemexpress LLC HY-105252A 5mg Medchemexpress, BF 227 CAS:845647-80-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY105252A5MG
Encompass_Preferred
Medchemexpress, HY-105252A 5mg BF 227 CAS:845647-80-5 BF 227 is a candidate for an amyloid imaging probe for PET, with a Ki of 4.3 nM for Aβ1-42 fibrils. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0683