Learn More
RWD LIFE SCIENCE INC C100-pro Automated Cell Counter C100-Pro, Bright Field and Flourencent Field, Cell Counting and Analysis in 9s, with FDA 21 CFR Part 11
SDP
Catalog No. NC3404390
Encompass_Preferred
A Real Time-Saver Speeds up Your ResearchRWD automated cell counter is a reliable cell counting device with highly intelligent analysis software and excellent microscopy optics structure.In order to meet the needs of cell therapy and biopharma, the data management and software system of C100-Pro fully conforms to FDA 21 CFR Part 11.
Catalog No. NC3404390 Supplier RWD LIFE SCIENCE INC Supplier No. C100PRO
Add to Cart
Add to Cart
Request a Quote
A Real Time-Saver Speeds up Your ResearchRWD automated cell counter is a reliable cell counting device with highly intelligent analysis software and excellent microscopy optics structure.In order to meet the needs of cell therapy and biopharma, the data management and software system of C100-Pro fully conforms to FDA 21 CFR Part 11.