Learn More
Medchemexpress LLC HY-U00426 5mg Medchemexpress, BF738735 CAS:1436383-95-7 Purity:>98%
SDP
Catalog No. 501934442
Encompass_Preferred
Medchemexpress, HY-U00426 5mg BF738735 CAS:1436383-95-7 BF738735 is a phosphatidylinositol 4-kinase III beta ( PI4KIIIβ ) inhibitor with an IC 50 of 5.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4442 Supplier Medchemexpress LLC Supplier No. HYU004265MG
Medchemexpress, HY-U00426 5mg BF738735 CAS:1436383-95-7 BF738735 is a phosphatidylinositol 4-kinase III beta ( PI4KIIIβ ) inhibitor with an IC 50 of 5.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.