Learn More
MSC BF7881 BOX-STYLE
Catalog No. 5000504565
Encompass
Box-style fuel filters protect sensitive fuel system components from damaging contaminants, such as injection pumps and injectors.
  • Product type: Box-style fuel filter with bail handle
  • Size: 2 31/32 x 2 31/32 x 2 1/8 in (75.4 x 75.4 x 54.0 mm)
Catalog No. 50-005-04565 Supplier MSC Supplier No. 10952703
Box-style fuel filters protect sensitive fuel system components from damaging contaminants, such as injection pumps and injectors.
  • Product type: Box-style fuel filter with bail handle
  • Size: 2 31/32 x 2 31/32 x 2 1/8 in (75.4 x 75.4 x 54.0 mm)