Learn More
MSC BF7912 BOX-STYLE
Catalog No. 5000504543
Encompass
Baldwin's box-style fuel filters protect sensitive fuel system components, like injection pumps and injectors, from damaging contaminants.
  • Box-style fuel element with bail handle
  • Gasket attached
  • Compatible with M.A.N. trucks and Volvo excavators
Catalog No. 50-005-04543 Supplier MSC Supplier No. 10952695
Baldwin's box-style fuel filters protect sensitive fuel system components, like injection pumps and injectors, from damaging contaminants.
  • Box-style fuel element with bail handle
  • Gasket attached
  • Compatible with M.A.N. trucks and Volvo excavators