Learn More
New England Biolabs, Inc. BfaI – 500 units
SDP
Catalog No. 50812259
Encompass
A restriction endonuclease that recognizes the sequence C^TA_G.
Catalog No. 50-812-259 Supplier New England Biolabs, Inc. Supplier No. R0568S
A restriction endonuclease that recognizes the sequence C^TA_G.