Learn More
Aviva Systems Biology Corp Bfar Antibody - middle region (ARP43221_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43221_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Bfar. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 3010001A07Rik, AI666707, AW107665, BAR, RNF47, Bar, Rnf47ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-064