Learn More
AdipoGen BFC
SDP
Catalog No. 502561606
Encompass_Preferred
Chemical. CAS 220001-53-6. Formula C17H11F3O3. MW 320.3. 7-Benzyloxy-4-trifluoromethylcoumarin is a fluorescent substrate for cytochrome P450 family members CYP1A2, CYP3A4 and potentially others. Has been used in high-throughput screens in fluorometric p450 activity assays and is metabolized to the fluorescent metabolite 7-Hydroxy-4-trifluoromethylcoumarin HFC with lambdaex=410nm lambdaem=530nm.
Catalog No. 50-256-1606 Supplier AdipoGen Supplier No. CDXB0299M250
Chemical. CAS 220001-53-6. Formula C17H11F3O3. MW 320.3. 7-Benzyloxy-4-trifluoromethylcoumarin is a fluorescent substrate for cytochrome P450 family members CYP1A2, CYP3A4 and potentially others. Has been used in high-throughput screens in fluorometric p450 activity assays and is metabolized to the fluorescent metabolite 7-Hydroxy-4-trifluoromethylcoumarin HFC with lambdaex=410nm lambdaem=530nm.