Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP549310UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: Untagged. Host: E. coli. Species Reactivity: Rat. BioActivity: BioActivity: Shows bioactivity at a concentration of 1 ng/mL in the BALB/c 3T3 proliferation assay. The bioactivity acceptance criteria is set at 1.0 x 10^6 units/mg. AA Sequence: MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS. Buffer: 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5. Purity: Greater than 95% as determined by SDS-PAGE. Endotoxin: Less than or equal to 1.00 EUs/ug as determined by the LAL method. Gene Symbol: FGF2. Gene ID: 54250. Uniprot: P13109.
Catalog No. 50-188-5863