Learn More
Medchemexpress LLC HY-100419 5mg Medchemexpress, BFH772 CAS:890128-81-1 Purity:>98%
SDP
Catalog No. 501967729
Encompass_Preferred
Medchemexpress, HY-100419 5mg BFH772 CAS:890128-81-1 BFH772 is a potent oral VEGFR2 inhibitor, which is highly effective at targeting VEGFR2 kinase with an IC50 value of 3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7729 Supplier Medchemexpress LLC Supplier No. HY1004195MG
Medchemexpress, HY-100419 5mg BFH772 CAS:890128-81-1 BFH772 is a potent oral VEGFR2 inhibitor, which is highly effective at targeting VEGFR2 kinase with an IC50 value of 3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.