Learn More
Cayman Chemical Bfl-1humnrecombInt 200ug
SDP
Catalog No. 502945094
Encompass_Preferred
Source: Recombinant human N-terminal His-tagged Bfl-1 expressed in E. coli • Amino acids: 1-152 • MW: 19.7 kDa
Catalog No. 50-294-5094 Supplier Cayman Chemical Supplier No. 41998200 UG
Source: Recombinant human N-terminal His-tagged Bfl-1 expressed in E. coli • Amino acids: 1-152 • MW: 19.7 kDa